Get a Camera and C-Mount Adapter When You Buy 4 Motic Microscopes
When you buy four Motic™ Panthera C2 Compound LED Microscopes, you’ll get a Motic Moticam X5 Plus camera and Motic 0.35X C-Mount Camera Adapter at no additional cost. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
The Panthera C2 microscope has advanced features such as the Motic LightTracer. The LightTracer function remembers your last Illumination level for each magnification and adjusts the level automatically to your last setting as you rotate the nosepiece. It also has a sleep function to save power after 15 minutes of nonuse. Additionally, it’s capable of phase contrast utilizing a phase slider or both phase contrast and darkfield microscopy with the phase turret condenser. Other options include a simple polarizer for polarization contrast and a ready-to-go LED fluorescence module for fluorescence microscopy.
The Moticam X5 Plus is a high-resolution streaming Wi-Fi camera you can connect to with your Android or iOS device and view, capture, or edit live images from your microscope with the MotiConnect app.
This C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
| Catalog Number | Description | ||
|---|---|---|---|
Motic Moticam X5 Plus Camera | Learn More | ||
Motic 0.35X C-Mount Camera Adapter | Learn More |