Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UGGUAGACUAUGGAACGUAGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-379-5p | MIMAT0000733 |
| Mouse | mmu-miR-379-5p | MIMAT0000743 |
| Rat | rno-miR-379-5p | MIMAT0003192 |
| Bovine | bta-miR-379 | MIMAT0009306 |
| Cricetulus griseus | cgr-miR-379 | MIMAT0023945 |
| Dasypus novemcinctus | dno-miR-379-5p | MIMAT0047860 |
| View More Rows | ||
| Dog | cfa-miR-379 | MIMAT0006715 |
| Goat | chi-miR-379-5p | MIMAT0036195 |
| Gorilla gorilla | ggo-miR-379 | MIMAT0024110 |
| Guinea pig | cpo-miR-379-5p | MIMAT0047221 |
| Horse | eca-miR-379 | MIMAT0013147 |
| Pan paniscus | ppa-miR-379 | MIMAT0049047 |
| Pan troglodytes | ptr-miR-379 | MIMAT0008131 |
| Pongo pygmaeus | ppy-miR-379 | MIMAT0015869 |
| Pteropus alecto | pal-miR-379-5p | MIMAT0039886 |
| Rabbit | ocu-miR-379-5p | MIMAT0048423 |
| Rhesus monkey | mml-miR-379-5p | MIMAT0006308 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Dasypus novemcinctus
Dog
Goat
Gorilla gorilla
Guinea pig
Horse
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map