Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UGGUAGACUAUGGAACGUAGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-379-5p | MIMAT0000733 | |
| Mouse | mmu-miR-379-5p | MIMAT0000743 | |
| Rat | rno-miR-379-5p | MIMAT0003192 | |
| Bovine | bta-miR-379 | MIMAT0009306 | |
| Cricetulus griseus | cgr-miR-379 | MIMAT0023945 | |
| Dasypus novemcinctus | dno-miR-379-5p | MIMAT0047860 | |
| View More Rows | |||
| Dog | cfa-miR-379 | MIMAT0006715 | |
| Goat | chi-miR-379-5p | MIMAT0036195 | |
| Gorilla gorilla | ggo-miR-379 | MIMAT0024110 | |
| Guinea pig | cpo-miR-379-5p | MIMAT0047221 | |
| Horse | eca-miR-379 | MIMAT0013147 | |
| Pan paniscus | ppa-miR-379 | MIMAT0049047 | |
| Pan troglodytes | ptr-miR-379 | MIMAT0008131 | |
| Pongo pygmaeus | ppy-miR-379 | MIMAT0015869 | |
| Pteropus alecto | pal-miR-379-5p | MIMAT0039886 | |
| Rabbit | ocu-miR-379-5p | MIMAT0048423 | |
| Rhesus monkey | mml-miR-379-5p | MIMAT0006308 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Cricetulus griseus
Dasypus novemcinctus
Dog
Goat
Gorilla gorilla
Guinea pig
Horse
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-379 (v17)
Mouse: mmu-miR-379 (v17)
Rat: rno-miR-379 (v18)
Macaca mulatta: mml-miR-379 (v19)
Mouse: mmu-miR-379 (v17)
Rat: rno-miR-379 (v18)
Macaca mulatta: mml-miR-379 (v19)

Genomic Map