Assay Details
Gene Family ID: | MIPF0000397, mir-708 |
Mature miRNA Sequence: | CAACUAGACUGUGAGCUUCUAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-708-3p | MIMAT0004927 |
| Mouse | mmu-miR-708-3p | MIMAT0003498 |
| Rat | rno-miR-708-3p | MIMAT0005332 |
Stem-loop Details
Human
Mouse
Rat
More Information
Related Products
miRBase Alias:
Human: hsa-miR-708* (v17)
Mouse: mmu-miR-708* (v17)
Rat: rno-miR-708* (v18)
Mouse: mmu-miR-708* (v17)
Rat: rno-miR-708* (v18)

Genomic Map