Assay Details
Amplicon Length: | 76 |
Chromosome Location: | Chr. 7 - 96249718 - 96249718 [+] on 96249718 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-708-3p | MIMAT0003498 |
Mouse | mmu-miR-708-5p | MIMAT0004828 |
Stem-loop Details
Mouse
