Assay Details
Amplicon Length: | 85 |
Chromosome Location: | Chr. 1 - 161221597 - 161221597 [+] on 161221597 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-708-3p | MIMAT0005332 |
| Rat | rno-miR-708-5p | MIMAT0005331 |
Stem-loop Details
Rat

Genomic Map