Assay Details
Amplicon Length: | 58 |
Chromosome Location: | Chr. 11 - 79401818 - 79401818 [-] on 79401818 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-708-3p | MIMAT0004927 |
| Human | hsa-miR-708-5p | MIMAT0004926 |
Stem-loop Details
Human

Genomic Map