Assay Details
Gene Family ID: | MIPF0000397, mir-708 |
Mature miRNA Sequence: | AAGGAGCUUACAAUCUAGCUGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-708-5p | MIMAT0004926 |
Mouse | mmu-miR-708-5p | MIMAT0004828 |
Rat | rno-miR-708-5p | MIMAT0005331 |
Bovine | bta-miR-708 | MIMAT0009367 |
Callithrix jacchus | cja-miR-708 | MIMAT0039351 |
Dasypus novemcinctus | dno-miR-708-5p | MIMAT0048006 |
View More Rows | ||
Dog | cfa-miR-708 | MIMAT0006643 |
Goat | chi-miR-708-5p | MIMAT0036290 |
Guinea pig | cpo-miR-708-5p | MIMAT0047377 |
Horse | eca-miR-708 | MIMAT0012993 |
Microcebus murinus | mmr-miR-708 | MIMAT0049137 |
Pan troglodytes | ptr-miR-708 | MIMAT0008315 |
Papio hamadryas | pha-miR-708 | MIMAT0049572 |
Pig | ssc-miR-708-5p | MIMAT0013945 |
Pongo pygmaeus | ppy-miR-708 | MIMAT0016100 |
Rabbit | ocu-miR-708-5p | MIMAT0048561 |
Rhesus monkey | mml-miR-708-5p | MIMAT0012785 |
Tupaia chinensis | tch-miR-708-5p | MIMAT0036587 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Microcebus murinus
Pan troglodytes
Papio hamadryas
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Tupaia chinensis

More Information
Related Products
miRBase Alias:
Human: hsa-miR-708 (v17)
Mouse: mmu-miR-708 (v17)
Rat: rno-miR-708 (v18)
Macaca mulatta: mml-miR-708 (v19)
Staphylococcus schleiferi: ssc-miR-708 (v15)
Mouse: mmu-miR-708 (v17)
Rat: rno-miR-708 (v18)
Macaca mulatta: mml-miR-708 (v19)
Staphylococcus schleiferi: ssc-miR-708 (v15)
