Assay Details
Gene Family ID: | MIPF0000397, mir-708 |
Mature miRNA Sequence: | AAGGAGCUUACAAUCUAGCUGGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-708-5p | MIMAT0004926 |
Mouse | mmu-miR-708-5p | MIMAT0004828 |
Rat | rno-miR-708-5p | MIMAT0005331 |
Bovine | bta-miR-708 | MIMAT0009367 |
Callithrix jacchus | cja-miR-708 | MIMAT0039351 |
Dasypus novemcinctus | dno-miR-708-5p | MIMAT0048006 |
View More Rows | ||
Dog | cfa-miR-708 | MIMAT0006643 |
Goat | chi-miR-708-5p | MIMAT0036290 |
Guinea pig | cpo-miR-708-5p | MIMAT0047377 |
Horse | eca-miR-708 | MIMAT0012993 |
Microcebus murinus | mmr-miR-708 | MIMAT0049137 |
Pan troglodytes | ptr-miR-708 | MIMAT0008315 |
Papio hamadryas | pha-miR-708 | MIMAT0049572 |
Pig | ssc-miR-708-5p | MIMAT0013945 |
Pongo pygmaeus | ppy-miR-708 | MIMAT0016100 |
Rabbit | ocu-miR-708-5p | MIMAT0048561 |
Rhesus monkey | mml-miR-708-5p | MIMAT0012785 |
Tupaia chinensis | tch-miR-708-5p | MIMAT0036587 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Microcebus murinus
Pan troglodytes
Papio hamadryas
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Tupaia chinensis
