Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | UGAUAUGUUUGAUAUAUUAGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-190a-5p | MIMAT0000458 |
| Mouse | mmu-miR-190a-5p | MIMAT0000220 |
| Rat | rno-miR-190a-5p | MIMAT0000865 |
| Bovine | bta-miR-190a | MIMAT0009251 |
| Chicken | gga-miR-190a-5p | MIMAT0001155 |
| Chrysemys picta | cpi-miR-190a-5p | MIMAT0037804 |
| View More Rows | ||
| Dog | cfa-miR-190a | MIMAT0009872 |
| Fugu rubripes | fru-miR-190 | MIMAT0003109 |
| Gorilla gorilla | ggo-miR-190a | MIMAT0002314 |
| Horse | eca-miR-190a | MIMAT0012896 |
| Monodelphis domestica | mdo-miR-190a-5p | MIMAT0012740 |
| Ornithorhynchus anatinus | oan-miR-190a-5p | MIMAT0006843 |
| Pan paniscus | ppa-miR-190 | MIMAT0002315 |
| Pan troglodytes | ptr-miR-190a | MIMAT0002313 |
| Pongo pygmaeus | ppy-miR-190a | MIMAT0015783 |
| Rhesus monkey | mml-miR-190a-5p | MIMAT0002312 |
| Sarcophilus harrisii | sha-miR-190 | MIMAT0022769 |
| Taeniopygia guttata | tgu-miR-190-5p | MIMAT0014589 |
| Tetraodon nigroviridis | tni-miR-190 | MIMAT0003110 |
| Zebrafish | dre-miR-190a | MIMAT0001854 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-190a |
| Stem-loop Accession # | MI0000486 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | Chr. 15 - 62823957 - 62824041 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000458
miRBase ID: hsa-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: Chr. 15 - 62823957 - 62824041 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-190a |
| Stem-loop Accession # | MI0000232 |
| Stem-loop Sequence | CUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAGCAUAUUCCUACAG |
| Chromosome Location | Chr. 9 - 67236660 - 67236726 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000220
miRBase ID: mmu-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: Chr. 9 - 67236660 - 67236726 [-] on Build GRCm38 |
| Stem-loop ID | rno-mir-190a-1 |
| Stem-loop Accession # | MI0000933 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAGCAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | Chr. 8 - 73030047 - 73030131 [-] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000865
miRBase ID: rno-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: Chr. 8 - 80928165 - 80928249 [+] on Build Rnor_6.0 |
| Stem-loop ID | rno-mir-190a-2 |
| Stem-loop Accession # | MI0031759 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAGCAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | Chr. 8 - 80928165 - 80928249 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000865
miRBase ID: rno-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: Chr. 8 - 80928165 - 80928249 [+] on Build Rnor_6.0 |
| Stem-loop ID | bta-mir-190a |
| Stem-loop Accession # | MI0009762 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009251
miRBase ID: bta-miR-190a Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | gga-mir-190a |
| Stem-loop Accession # | MI0001224 |
| Stem-loop Sequence | UGCAGGACUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCCUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001155
miRBase ID: gga-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | cpi-mir-190a |
| Stem-loop Accession # | MI0029510 |
| Stem-loop Sequence | UGCAGGGCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUAUAGUGUCCUGCCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037804
miRBase ID: cpi-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | cfa-mir-190a |
| Stem-loop Accession # | MI0010369 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009872
miRBase ID: cfa-miR-190a Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | fru-mir-190 |
| Stem-loop Accession # | MI0003473 |
| Stem-loop Sequence | CUGUCUGAUAUGUUUGAUAUAUUAGGUUGUUAUUCAGUCCAACUAUAUAUCAAACAUAUUCCUACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003109
miRBase ID: fru-miR-190 Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | ggo-mir-190a |
| Stem-loop Accession # | MI0002615 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002314
miRBase ID: ggo-miR-190a Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | eca-mir-190a |
| Stem-loop Accession # | MI0012652 |
| Stem-loop Sequence | CUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012896
miRBase ID: eca-miR-190a Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | mdo-mir-190a |
| Stem-loop Accession # | MI0012496 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCCUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012740
miRBase ID: mdo-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | oan-mir-190a |
| Stem-loop Accession # | MI0006690 |
| Stem-loop Sequence | AGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006843
miRBase ID: oan-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | ppa-mir-190 |
| Stem-loop Accession # | MI0002616 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002315
miRBase ID: ppa-miR-190 Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | ptr-mir-190a |
| Stem-loop Accession # | MI0002614 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002313
miRBase ID: ptr-miR-190a Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | ppy-mir-190a |
| Stem-loop Accession # | MI0014851 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015783
miRBase ID: ppy-miR-190a Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | mml-mir-190a |
| Stem-loop Accession # | MI0002613 |
| Stem-loop Sequence | UGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCUUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002312
miRBase ID: mml-miR-190a-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | sha-mir-190 |
| Stem-loop Accession # | MI0019595 |
| Stem-loop Sequence | ACCUGGAUGCCUUUUCUGCAGGCCUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCCUGCCCUGUCUCCGGGGGUUCAUAAUAUAGUCUAUAAGGCAACAGGGAGGAGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022769
miRBase ID: sha-miR-190 Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | tgu-mir-190 |
| Stem-loop Accession # | MI0013817 |
| Stem-loop Sequence | AGACUCUGUGUGAUAUGUUUGAUAUAUUAGGUUGUUAUUUAAUCCAACUAUAUAUCAAACAUAUUCCUACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0014589
miRBase ID: tgu-miR-190-5p Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | tni-mir-190 |
| Stem-loop Accession # | MI0003474 |
| Stem-loop Sequence | CUGUCUGAUAUGUUUGAUAUAUUAGGUUGUUAUUCAGUCCAACUAUAUAUCAAACAUAUUCCUACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003110
miRBase ID: tni-miR-190 Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
| Stem-loop ID | dre-mir-190a |
| Stem-loop Accession # | MI0002027 |
| Stem-loop Sequence | UCUGGAGGUGAGGUAGACCUGGAAGCCUUUCUGCAGGCCUCUGUUUGAUAUGUUUGAUAUAUUAGGUUGUUAUUCUGUCCAACUAUAUAUCAAACAUAUUCCUACAGUGUCCUGCUCUGUCUCCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001854
miRBase ID: dre-miR-190a Mature miRNA Sequence: UGAUAUGUUUGAUAUAUUAGGU Chromosome Location: NA |
More Information
Related Products

Genomic Map