Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | UGAUAUGUUUGAUAUAUUAGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-190a-5p | MIMAT0000458 |
| Mouse | mmu-miR-190a-5p | MIMAT0000220 |
| Rat | rno-miR-190a-5p | MIMAT0000865 |
| Bovine | bta-miR-190a | MIMAT0009251 |
| Chicken | gga-miR-190a-5p | MIMAT0001155 |
| Chrysemys picta | cpi-miR-190a-5p | MIMAT0037804 |
| View More Rows | ||
| Dog | cfa-miR-190a | MIMAT0009872 |
| Fugu rubripes | fru-miR-190 | MIMAT0003109 |
| Gorilla gorilla | ggo-miR-190a | MIMAT0002314 |
| Horse | eca-miR-190a | MIMAT0012896 |
| Monodelphis domestica | mdo-miR-190a-5p | MIMAT0012740 |
| Ornithorhynchus anatinus | oan-miR-190a-5p | MIMAT0006843 |
| Pan paniscus | ppa-miR-190 | MIMAT0002315 |
| Pan troglodytes | ptr-miR-190a | MIMAT0002313 |
| Pongo pygmaeus | ppy-miR-190a | MIMAT0015783 |
| Rhesus monkey | mml-miR-190a-5p | MIMAT0002312 |
| Sarcophilus harrisii | sha-miR-190 | MIMAT0022769 |
| Taeniopygia guttata | tgu-miR-190-5p | MIMAT0014589 |
| Tetraodon nigroviridis | tni-miR-190 | MIMAT0003110 |
| Zebrafish | dre-miR-190a | MIMAT0001854 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Chicken
Chrysemys picta
Dog
Fugu rubripes
Gorilla gorilla
Horse
Monodelphis domestica
Ornithorhynchus anatinus
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Sarcophilus harrisii
Taeniopygia guttata
Tetraodon nigroviridis
Zebrafish

Genomic Map