Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 12 - 109735129 - 109735129 [+] on 109735129 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-485-5p | MIMAT0003128 |
| Mouse | mmu-miR-485-3p | MIMAT0003129 |
Stem-loop Details
Mouse

Genomic Map