Assay Details
Gene Family ID: | MIPF0000201, mir-485 |
Mature miRNA Sequence: | AGAGGCUGGCCGUGAUGAAUUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-485-5p | MIMAT0002175 | |
| Mouse | mmu-miR-485-5p | MIMAT0003128 | |
| Rat | rno-miR-485-5p | MIMAT0003203 | |
| Dasypus novemcinctus | dno-miR-485-5p | MIMAT0047910 | |
| Goat | chi-miR-485-5p | MIMAT0036244 | |
| Horse | eca-miR-485-5p | MIMAT0013159 | |
| View More Rows | |||
| Pan paniscus | ppa-miR-485 | MIMAT0049037 | |
| Pongo pygmaeus | ppy-miR-485-5p | MIMAT0015902 | |
| Pteropus alecto | pal-miR-485-5p | MIMAT0039878 | |
| Rhesus monkey | mml-miR-485-5p | MIMAT0006342 | |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Goat
Horse
Pan paniscus
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool B v3.0
Related Products
miRBase Alias:
Mouse: mmu-miR-485 (v17)
Rat: rno-miR-485 (v18)
Rat: rno-miR-485 (v18)

Genomic Map