Assay Details
Gene Family ID: | MIPF0000201, mir-485 |
Mature miRNA Sequence: | AGAGGCUGGCCGUGAUGAAUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-485-5p | MIMAT0002175 |
| Mouse | mmu-miR-485-5p | MIMAT0003128 |
| Rat | rno-miR-485-5p | MIMAT0003203 |
| Dasypus novemcinctus | dno-miR-485-5p | MIMAT0047910 |
| Goat | chi-miR-485-5p | MIMAT0036244 |
| Horse | eca-miR-485-5p | MIMAT0013159 |
| View More Rows | ||
| Pan paniscus | ppa-miR-485 | MIMAT0049037 |
| Pongo pygmaeus | ppy-miR-485-5p | MIMAT0015902 |
| Pteropus alecto | pal-miR-485-5p | MIMAT0039878 |
| Rhesus monkey | mml-miR-485-5p | MIMAT0006342 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Goat
Horse
Pan paniscus
Pongo pygmaeus
Pteropus alecto
Rhesus monkey

Genomic Map