Assay Details
Gene Family ID: | MIPF0000201, mir-485 |
Mature miRNA Sequence: | AGAGGCUGGCCGUGAUGAAUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-485-5p | MIMAT0002175 |
Mouse | mmu-miR-485-5p | MIMAT0003128 |
Rat | rno-miR-485-5p | MIMAT0003203 |
Dasypus novemcinctus | dno-miR-485-5p | MIMAT0047910 |
Goat | chi-miR-485-5p | MIMAT0036244 |
Horse | eca-miR-485-5p | MIMAT0013159 |
View More Rows | ||
Pan paniscus | ppa-miR-485 | MIMAT0049037 |
Pongo pygmaeus | ppy-miR-485-5p | MIMAT0015902 |
Pteropus alecto | pal-miR-485-5p | MIMAT0039878 |
Rhesus monkey | mml-miR-485-5p | MIMAT0006342 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Goat
Horse
Pan paniscus
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
