Assay Details
Amplicon Length: | 58 |
Chromosome Location: | Chr. 12 - 109718551 - 109718551 [-] on 109718551 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-495-5p | MIMAT0017249 |
| Mouse | mmu-miR-495-3p | MIMAT0003456 |
Stem-loop Details
Mouse

Genomic Map