Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AAACAAACAUGGUGCACUUCUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-495-3p | MIMAT0002817 | |
| Mouse | mmu-miR-495-3p | MIMAT0003456 | |
| Rat | rno-miR-495 | MIMAT0005320 | |
| Bovine | bta-miR-495 | MIMAT0009335 | |
| Callithrix jacchus | cja-miR-495 | MIMAT0039338 | |
| Dasypus novemcinctus | dno-miR-495-3p | MIMAT0047931 | |
| View More Rows | |||
| Dog | cfa-miR-495 | MIMAT0006721 | |
| Goat | chi-miR-495-3p | MIMAT0036256 | |
| Guinea pig | cpo-miR-495-3p | MIMAT0047292 | |
| Horse | eca-miR-495 | MIMAT0013165 | |
| Pan troglodytes | ptr-miR-495 | MIMAT0008169 | |
| Pongo pygmaeus | ppy-miR-495 | MIMAT0015916 | |
| Pteropus alecto | pal-miR-495-3p | MIMAT0040186 | |
| Rabbit | ocu-miR-495-3p | MIMAT0048484 | |
| Rhesus monkey | mml-miR-495-3p | MIMAT0006357 | |
| Sheep | oar-miR-495-3p | MIMAT0019274 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Sheep
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-495 (v18)
Mouse: mmu-miR-495 (v17)
Macaca mulatta: mml-miR-495 (v19)
Mouse: mmu-miR-495 (v17)
Macaca mulatta: mml-miR-495 (v19)

Genomic Map