Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AAACAAACAUGGUGCACUUCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-495-3p | MIMAT0002817 |
| Mouse | mmu-miR-495-3p | MIMAT0003456 |
| Rat | rno-miR-495 | MIMAT0005320 |
| Bovine | bta-miR-495 | MIMAT0009335 |
| Callithrix jacchus | cja-miR-495 | MIMAT0039338 |
| Dasypus novemcinctus | dno-miR-495-3p | MIMAT0047931 |
| View More Rows | ||
| Dog | cfa-miR-495 | MIMAT0006721 |
| Goat | chi-miR-495-3p | MIMAT0036256 |
| Guinea pig | cpo-miR-495-3p | MIMAT0047292 |
| Horse | eca-miR-495 | MIMAT0013165 |
| Pan troglodytes | ptr-miR-495 | MIMAT0008169 |
| Pongo pygmaeus | ppy-miR-495 | MIMAT0015916 |
| Pteropus alecto | pal-miR-495-3p | MIMAT0040186 |
| Rabbit | ocu-miR-495-3p | MIMAT0048484 |
| Rhesus monkey | mml-miR-495-3p | MIMAT0006357 |
| Sheep | oar-miR-495-3p | MIMAT0019274 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Sheep

Genomic Map