Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AAACAAACAUGGUGCACUUCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-495-3p | MIMAT0002817 |
| Mouse | mmu-miR-495-3p | MIMAT0003456 |
| Rat | rno-miR-495 | MIMAT0005320 |
| Bovine | bta-miR-495 | MIMAT0009335 |
| Callithrix jacchus | cja-miR-495 | MIMAT0039338 |
| Dasypus novemcinctus | dno-miR-495-3p | MIMAT0047931 |
| View More Rows | ||
| Dog | cfa-miR-495 | MIMAT0006721 |
| Goat | chi-miR-495-3p | MIMAT0036256 |
| Guinea pig | cpo-miR-495-3p | MIMAT0047292 |
| Horse | eca-miR-495 | MIMAT0013165 |
| Pan troglodytes | ptr-miR-495 | MIMAT0008169 |
| Pongo pygmaeus | ppy-miR-495 | MIMAT0015916 |
| Pteropus alecto | pal-miR-495-3p | MIMAT0040186 |
| Rabbit | ocu-miR-495-3p | MIMAT0048484 |
| Rhesus monkey | mml-miR-495-3p | MIMAT0006357 |
| Sheep | oar-miR-495-3p | MIMAT0019274 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Sheep
More Information
Related Products
miRBase Alias:
Human: hsa-miR-495 (v18)
Mouse: mmu-miR-495 (v17)
Macaca mulatta: mml-miR-495 (v19)
Mouse: mmu-miR-495 (v17)
Macaca mulatta: mml-miR-495 (v19)

Genomic Map