Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 13 - 58023418 - 58023418 [+] on 58023418 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-874-3p | MIMAT0004853 |
Mouse | mmu-miR-874-5p | MIMAT0017268 |
Stem-loop Details
Mouse
