Assay Details
Gene Family ID: | MIPF0000401, mir-874 |
Mature miRNA Sequence: | CUGCCCUGGCCCGAGGGACCGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-874-3p | MIMAT0004911 |
Mouse | mmu-miR-874-3p | MIMAT0004853 |
Rat | rno-miR-874-3p | MIMAT0005284 |
Bovine | bta-miR-874 | MIMAT0009378 |
Dog | cfa-miR-874 | MIMAT0009930 |
Gorilla gorilla | ggo-miR-874 | MIMAT0024140 |
View More Rows | ||
Horse | eca-miR-874 | MIMAT0013069 |
Pan troglodytes | ptr-miR-874 | MIMAT0008325 |
Pongo pygmaeus | ppy-miR-874 | MIMAT0016112 |
Rhesus monkey | mml-miR-874-3p | MIMAT0006516 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Gorilla gorilla
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-874 (v19)
Mouse: mmu-miR-874 (v17)
Rat: rno-miR-874 (v18)
Macaca mulatta: mml-miR-874 (v19)
Mouse: mmu-miR-874 (v17)
Rat: rno-miR-874 (v18)
Macaca mulatta: mml-miR-874 (v19)
