Assay Details
Gene Family ID: | MIPF0000401, mir-874 |
Mature miRNA Sequence: | CUGCCCUGGCCCGAGGGACCGA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-874-3p | MIMAT0004911 | |
Mouse | mmu-miR-874-3p | MIMAT0004853 | |
Rat | rno-miR-874-3p | MIMAT0005284 | |
Bovine | bta-miR-874 | MIMAT0009378 | |
Dog | cfa-miR-874 | MIMAT0009930 | |
Gorilla gorilla | ggo-miR-874 | MIMAT0024140 | |
View More Rows | |||
Horse | eca-miR-874 | MIMAT0013069 | |
Pan troglodytes | ptr-miR-874 | MIMAT0008325 | |
Pongo pygmaeus | ppy-miR-874 | MIMAT0016112 | |
Rhesus monkey | mml-miR-874-3p | MIMAT0006516 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Gorilla gorilla
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-874 (v19)
Mouse: mmu-miR-874 (v17)
Rat: rno-miR-874 (v18)
Macaca mulatta: mml-miR-874 (v19)
Mouse: mmu-miR-874 (v17)
Rat: rno-miR-874 (v18)
Macaca mulatta: mml-miR-874 (v19)
