Assay Details
Gene Family ID: | MIPF0000401, mir-874 |
Mature miRNA Sequence: | CUGCCCUGGCCCGAGGGACCGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-874-3p | MIMAT0004911 |
| Mouse | mmu-miR-874-3p | MIMAT0004853 |
| Rat | rno-miR-874-3p | MIMAT0005284 |
| Bovine | bta-miR-874 | MIMAT0009378 |
| Dog | cfa-miR-874 | MIMAT0009930 |
| Gorilla gorilla | ggo-miR-874 | MIMAT0024140 |
| View More Rows | ||
| Horse | eca-miR-874 | MIMAT0013069 |
| Pan troglodytes | ptr-miR-874 | MIMAT0008325 |
| Pongo pygmaeus | ppy-miR-874 | MIMAT0016112 |
| Rhesus monkey | mml-miR-874-3p | MIMAT0006516 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Gorilla gorilla
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map