Assay Details
Gene Family ID: | MIPF0000162, mir-367 |
Mature miRNA Sequence: | AAUUGCACUUUAGCAAUGGUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-367-3p | MIMAT0000719 |
| Mouse | mmu-miR-367-3p | MIMAT0003181 |
| Anolis carolinensis | aca-miR-367 | MIMAT0021949 |
| Columba livia | cli-miR-367-3p | MIMAT0038627 |
| Dasypus novemcinctus | dno-miR-367-3p | MIMAT0047839 |
| Guinea pig | cpo-miR-367-3p | MIMAT0047198 |
| View More Rows | ||
| Horse | eca-miR-367 | MIMAT0012918 |
| Pan troglodytes | ptr-miR-367 | MIMAT0008118 |
| Pongo pygmaeus | ppy-miR-367 | MIMAT0015854 |
| Rabbit | ocu-miR-367-3p | MIMAT0036325 |
| Rhesus monkey | mml-miR-367 | MIMAT0006292 |
| Taeniopygia guttata | tgu-miR-367 | MIMAT0025395 |
Stem-loop Details
Human
Mouse
Anolis carolinensis
Columba livia
Dasypus novemcinctus
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Human: hsa-miR-367 (v17)
Mouse: mmu-miR-367 (v17)
Mouse: mmu-miR-367 (v17)

Genomic Map