Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 6 - 133885148 - 133885148 [-] on 133885148 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-485-5p | MIMAT0003203 |
Rat | rno-miR-485-3p | MIMAT0017222 |
Stem-loop Details
Rat
