Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 17 - 7025465 - 7025465 [-] on 7025465 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-874-3p | MIMAT0005284 |
| Rat | rno-miR-874-5p | MIMAT0017290 |
Stem-loop Details
Rat

Genomic Map