Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,3-Trimethoxypropane, 97%

Catalog No. p-7027013
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2344814

Supplier: Thermo Scientific Chemicals B2344814

Only null left
Add to Cart

CAS: 14315-97-0 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14315-97-0
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008490
InChI Key FKZYYYDRLJCHGL-UHFFFAOYSA-N
Synonym propane, 1,1,3-trimethoxy, propane,1,1,3-trimethoxy, acmc-1brb6, 1,1,3-trimethoxy-propane, 1,1,3-trimethoxypropane, beta-methoxypropionaldehyde dimethyl acetal
PubChem CID 84348
IUPAC Name 1,1,3-trimethoxypropane
SMILES COCCC(OC)OC

Specifications

Density 0.942
Boiling Point 45°C to 46°C (17 mmHg)
Flash Point 40°C (105°F)
Refractive Index 1.4
Quantity 25 g
UN Number UN3271
Formula Weight 134.17
Percent Purity 98%
Chemical Name or Material 1,1,3-Trimethoxypropane
Hazard Category H226
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
RTECSNumber TZ9560000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.