Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorobenzyl)piperazine, 98%

Catalog No. p-7052691
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. AAH5533703

Supplier: Thermo Scientific Chemicals H5533703

Only null left
Add to Cart

CAS: 23145-88-2 | C11H15ClN2 | 210.705 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23145-88-2
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.705
MDL Number MFCD00040791
InChI Key GSJXJZOWHSTWOX-UHFFFAOYSA-N
Synonym 1-4-chlorobenzyl piperazine, 1-4-chlorophenyl methyl piperazine, 4-chlorobenzylpiperazine, 1-4-chlorobenzyl-piperazine, 1-4-chloro-benzyl-piperazine, 1-4-chloro benzyl piperazine, n-4-chlorobenzyl piperazine, 4-chlorophenyl methyl piperazine, norhomochlorcyclizine, chlorobenzylpiperazin
PubChem CID 134826
IUPAC Name 1-[(4-chlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)Cl

Specifications

Color Colorless to Yellow
Density 1.140 g/mL
Boiling Point 104°C to 106°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.558
Quantity 1 g
UN Number UN3267
Formula Weight 210.71
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(4-Chlorobenzyl)piperazine
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.