Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T1516500G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 433-06-7
Molecular Formula C9H9F3O3S
Molecular Weight (g/mol) 254.223
MDL Number MFCD00000443
InChI Key IGKCQDUYZULGBM-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl p-toluenesulfonate, 2,2,2-trifluoroethyl tosylate, trifluoroethanol tosylate, trifluorethyltosylate, 2,2,2-trifluoroethyl-p-toluenesulfonate, ethanol, 2,2,2-trifluoro-, 4-methylbenzenesulfonate, 2,2,2-trifluoroethyl p-toluenesulphonate, ethanol, 2,2,2-trifluoro-, p-toluenesulfonate, 2,2,2-trifluoroethyl 4-methylbenzene-1-sulfonate, ethanol, 2,2,2-trifluoro-, 1-4-methylbenzenesulfonate
PubChem CID 67942
IUPAC Name 2,2,2-trifluoroethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)F

Specifications

Melting Point 40°C
Color White
Boiling Point 104°C
Quantity 500 g
Formula Weight 254.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,2-Trifluoroethyl p-Toluenesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.