Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbutane, 99%

Catalog No. p-7026293
Click to view available options
Quantity:
25 g
100 g

CAS: 79-29-8 | C6H14 | 86.178 g/mol

2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Soluble in water (1mg/ml @22°C), DMSO, ethanol, acetone.

Notes
Incompatible with strong oxidizing agents, oxygen, ammonia.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C

Specifications

Melting Point -129°C
Density 0.662
Boiling Point 57°C to 59°C
Flash Point −29°C (−20°F)
Odor Mild
Refractive Index 1.375
Quantity 25 g
UN Number UN2457
Beilstein 1730737
Solubility Information Soluble in water (1mg/ml @22°C),DMSO,ethanol,acetone.
Formula Weight 86.18
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylbutane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2,3-DIMETHYLBUTANE
EINECSNumber 201-193-6
RTECSNumber EJ9350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.