Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chloroethyl methyl ether, 98%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.
Solubility
Soluble in water 60 g/L at 20°C.
Notes
Store away from oxidizing agents, bases.
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.
Solubility
Soluble in water 60 g/L at 20°C.
Notes
Store away from oxidizing agents, bases.
Specifications
Specifications
| Density | 1.04 |
| Boiling Point | 89°C to 90°C |
| Flash Point | 8°C (46°F) |
| Refractive Index | 1.409 |
| Linear Formula | CH3OCH2CH2Cl |
| Quantity | 25 g |
| UN Number | UN3271 |
| Solubility Information | Soluble in water 60g/L at 20°C. |
| Formula Weight | 94.54 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H225-H302 |
| Hazard Statement | GHS H Statement H225-H302-H332 Highly flammable liquid and vapour. Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P312-P330-P363-P370+P378q-P372-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S. |
| EINECSNumber | 210-999-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction