Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxyethyl Ether, 98+%, Extra Pure

Catalog No. p-3736417
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Metal drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117905000 500 g Glass bottle
AC117900010 1 kg Glass bottle
AC117900025 2.5 kg Metal drum
AC117900100 10 kg Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC117905000 Supplier Thermo Scientific Chemicals Supplier No. 117905000
Only null left
Add to Cart

CAS: 112-36-7 | C8H18O3 | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-36-7
MDL Number MFCD00009254
InChI Key RRQYJINTUHWNHW-UHFFFAOYSA-N
Synonym diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
PubChem CID 8179
ChEBI CHEBI:44664
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
SMILES CCOCCOCCOCC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Density 0.9g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C8H18O3
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Linear Formula (C2H5OCH2CH2)2O
Quantity 500 g
Residue after Evaporation 0.05% max.
Beilstein 01, II, 519
Merck Index 15, 3132
Molecular Weight (g/mol) 162.229
Formula Weight 162.23
Percent Purity ≥98.0%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl ether
Show More Show Less
Hazard Statement GHS H Statement:
May form explosive peroxides.
Precautionary Statement GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.