Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopyrimidine, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1450-85-7
C4H4N2S
112.15
MFCD00006073
HBCQSNAFLVXVAY-UHFFFAOYSA-N
2-mercaptopyrimidine,pyrimidine-2-thiol,2-pyrimidinethiol,2-thiopyrimidine,pyrimidine-2 1h-thione,2 1h-pyrimidinethione,pyrimidinethione,2-pyrimidinethione,2-pyrimidinethiol 9ci,2-1h-pyrimidinthione
1550489
CHEBI:53576
1H-pyrimidine-2-thione
C1=CNC(=S)N=C1
1450-85-7
C4H4N2S
112.15
MFCD00006073
HBCQSNAFLVXVAY-UHFFFAOYSA-N
2-mercaptopyrimidine,pyrimidine-2-thiol,2-pyrimidinethiol,2-thiopyrimidine,pyrimidine-2 1h-thione,2 1h-pyrimidinethione,pyrimidinethione,2-pyrimidinethione,2-pyrimidinethiol 9ci,2-1h-pyrimidinthione
1550489
CHEBI:53576
1H-pyrimidine-2-thione
C1=CNC(=S)N=C1
Melting Point 230°C
Color Yellow
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 112.15
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyrimidine, 98%
Show More Show Less

EINECSNumber : 215-917-3

TSCA : TSCA