Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Methylbutane, BAKER™, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Aluminum Bottle
Specifications
Specifications
| Flash Point | −51.00°C |
| Packaging | Aluminum Bottle |
| Quantity | 500 mL |
| Specific Gravity | 1L = 0.62kg |
| Formula Weight | 72.15 |
| Chemical Name or Material | 2-Methylbutane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction