Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Formylbenzo-15-crown 5-Ether 98.0+%, TCI America™
SDP

Catalog No. F0448100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 60835-73-6
Molecular Formula C15H20O6
Molecular Weight (g/mol) 296.319
MDL Number MFCD00192180
InChI Key MBJIKIAWNPEHOR-UHFFFAOYSA-N
Synonym 2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
PubChem CID 324243
IUPAC Name 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carbaldehyde
SMILES C1COCCOC2=C(C=C(C=C2)C=O)OCCOCCO1

Specifications

Melting Point 85°C
Quantity 100 mg
Formula Weight 296.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Formylbenzo-15-crown 5-Ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.