Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(thien-2-ylmethyl)benzylamine hydrochloride, 97%, Thermo Scientific™

Catalog No. CC70663CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC70663CB 250 mg Amber Glass Bottle
CC70663DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC70663CB Supplier Thermo Scientific Chemicals Supplier No. CC70663CB
Only null left
Add to Cart

Chemical Identifiers

CAS 1112459-82-1
Molecular Formula C12H14ClNS
Molecular Weight (g/mol) 239.761
MDL Number MFCD12198118
InChI Key BRDMSBXGZSSVJT-UHFFFAOYSA-N
Synonym 4-thien-2-ylmethyl benzylamine hydrochloride, 4-thiophen-2-ylmethyl phenyl methanamine hydrochloride, 4-2-thienylmethyl phenyl methylamine, chloride, 4-thien-2-ylmethyl phenyl methylamine hydrochloride, 1-4-thiophen-2-ylmethyl phenyl methanamine hydrochloride, 1-4-thiophen-2-yl methyl phenyl methanamine-hydrogen chloride 1/1
PubChem CID 43811053
IUPAC Name [4-(thiophen-2-ylmethyl)phenyl]methanamine;hydrochloride
SMILES C1=CSC(=C1)CC2=CC=C(C=C2)CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 239.77
Percent Purity 97%
Chemical Name or Material 4-(Thien-2-ylmethyl)benzylamine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.