Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 3A, powder <50 micron

Catalog No. AC214790025
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Color Undesignated
pH 11.4 (10% max. aq. suspension)
Quantity 2.5 kg
Packaging Plastic Drum
Particle Size 500 Micron Max.
Residual Water < 1.5% (550°C, 2 hrs.)
Water Adsorption > 22% (50%rH, 20°C, 24h)
Physical Form Powder or Crystals
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Molecular sieves 3A
Hazard Category HYGROSCOPIC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.