Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodouracil 99.0+%, TCI America™
SDP

Catalog No. I021925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 696-07-1
Molecular Formula C4H3IN2O2
Molecular Weight (g/mol) 237.98
MDL Number MFCD00006020
InChI Key KSNXJLQDQOIRIP-UHFFFAOYSA-N
Synonym 5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 69672
ChEBI CHEBI:43636
IUPAC Name 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES IC1=CNC(=O)NC1=O

Specifications

Color White
Quantity 25 g
Formula Weight 237.98
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodouracil
RTECSNumber YR0525000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.