Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ ECL™ start Western Blotting Detection Reagent
GSA_VA

Catalog No. p-5045893 Shop All Cytiva Products
Click to view available options
Quantity:
200 mL
400 mL

Reagent is luminol-based chemiluminescent substrate designed for the detection of proteins immobilized to a membrane

Amersham™ ECL™ start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins. Amersham ECL start has a long signal duration, which enables multiple exposures of the membrane plus a convenient time window between experiment and analysis.

  • Long signal duration provides assay design flexibility and robust detection of large experimental sets
  • Produces low background, regardless of method of detection
  • Working solution is stable for five days
  • Long shelf life of 18 hours is convenient for those who run Western blots infrequently
  • Detection on X-ray film or CCD Imager
  • Recommended Membrane: PVDF or nitrocellulose
  • For detection of high abundant proteins
  • Colorless detection reagent 1 and 2

Applications:

Routine and confirmatory Western blotting

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type ECL Western Blotting Detection Reagent
Quantity 200 mL
For Use With (Application) Routine and confirmatory Western blotting
Storage Requirements 18°C to 30°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.