Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxalase I Antibody (Glo1a), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$206.50 - $499.50
Specifications
| Antigen | Glyoxalase I |
|---|---|
| Clone | Glo1a |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Simple Western 1:50, ELISA 0.15 ng/ 1 ug protein, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature (10.3892/ol.2021.12808) |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP119015SS
![]() |
Novus Biologicals
NBP119015SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP119015
![]() |
Novus Biologicals
NBP119015 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Human, Mouse samples. It is validated for Western Blot, Simple Western, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Glyoxalase I | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| Q04760 | |
| 2739 | |
| Full length human GLO1 protein [Swiss-Prot #Q04760]. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Glo1a | |
| Western Blot, Simple Western 1:50, ELISA 0.15 ng/ 1 ug protein, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin reported in scientific literature (10.3892/ol.2021.12808) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
| GLO1 | |
| IgG1 κ | |
| Protein G purified | |
| 21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title