Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6-Chloro-7-deazapurine, 98%
Click to view available options
Quantity:
1 g
5 g
Description
6-Chloro-7-deazapurine is used for making tofacitinib.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Chloro-7-deazapurine is used for making tofacitinib.
Solubility
Soluble in DMSO, ethyl acetate and methanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
6-Chloro-7-deazapurine is used for making tofacitinib.
Solubility
Soluble in DMSO, ethyl acetate and methanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 190°C to 192°C |
| Quantity | 5 g |
| Solubility Information | Soluble in DMSO,ethyl acetate and methanol. |
| Formula Weight | 153.52 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Chloro-7-deazapurine |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction