Learn More
6-Chloropurine, 99%
Description
6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Chloropurine is used in the preparation of 9-alkylpurines through alkylation with various substituted alkyl halides in dimethyl sulfoxide. It is also used to prepare 6-succinoaminopurine.
Solubility
Soluble in hot water and dimethyl formamide.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 5774 |
| Merck Index | 14,2161 |
| Solubility Information | Soluble in hot water and dimethyl formamide. |
| Formula Weight | 154.56 |
| Percent Purity | 99% |
| Chemical Name or Material | 6-Chloropurine |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 201-745-6 |
| RTECSNumber | UO7520000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.