Learn More
Promega Rapid Digestion Kit---Trypsin/Lys-C, 100microg
Catalog No. prva1061 Shop All Promega Corporation Products
Click to view available options
Quantity:
100 μg
The Rapid Trypsin and Rapid Trypsin/Lys-C Digestion Kits are designed to shorten protein digestion times to 60 minutes versus the typical 4-18 hours.
  • Minimize Protein Digestion Time, Maximize Results
  • Faster digestion time
  • Streamlined workflow
  • Tighter coefficients of variation
Specifications
Content And Storage -30°C to -10°C
Quantity 100 μg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.