Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ A37

Click to view available options
Quantity:
10 mg
50 mg
Description
ALDH1A1 inhibitor (IC50 = 4.6 μM; Ki = 300 nM). Exhibits selectivity for ALDH1A1 over eight other ALDH isoforms. Disrupts spheroid formation and reduces cell viability of ovarian cancer cells in vitro.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 431.57 |
| Physical Form | Solid |
| Purity | >97% |
| Molecular Weight (g/mol) | 431.569 |
| Chemical Name or Material | Ethyl 2-[[3,4-dihydro-4-oxo-3-[3-(1-pyrrolidinyl)propyl][1]benzothieno[3,2-d]pyrimidin-2-yl]thio]ace |
Safety and Handling
| Recommended Storage | Soluble to 100mM in DMSO and to 100mM in ethanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction