Learn More
Description
Specifications
Specifications
| Antigen | ABCA1 |
| Applications | Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), KnockDown, Neutralization |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 19515742), Flow Cytometry 1:400. Use reported in scientific literature (PMID 21501868), ELISA reported in scientific literature (PMID 18541924), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100. Use reported in scientific literature (PMID 21501868), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 21846716; 21106520), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 27599291), Gel Super Shift Assays reported in scientific literature (PMID 15684432), Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 32273567), KnockDown Validated reported in scientific literature (PMID 31666189), Block/Neutralize reported in scientific literature (PMID 30821416), PCR reported in scientific literature (PMID 27406916), Dual RNAscope ISH-IHC |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O95477 |
| Gene Alias | ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1 |
| Gene Symbols | ABCA1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
