Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ABsolute Blue QPCR Mix, SYBR Green, low ROX

Catalog No. ab4322b
Click to view available options
No. of Reactions:
1600 Reactions
400 Reactions

Catalog No. AB4322B

Supplier: Thermo Scientific™ AB4322B

Only null left
Add to Cart
Add to Cart

Incorporates an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with ROX or Fluorescein passive reference dyes
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 16 x 1.25 mL ABsolute Blue QPCR SYBR Green Low ROX Mix (2X)
• 100 μL MgCl2 (1M)
Sufficient for 1600 x 25 μL reactions.

Store at -20°C
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
Product Line ABsolute Blue
Product Type Real Time PCR SYBR Master Mix
Quantity 1,600 reactions
Sufficient For 1600 Reactions of 25 μL Each
For Use With (Application) Gene Expression, Genotyping, siRNA Validation
Compatibility qPCR Cyclers
No. of Reactions 1600 Reactions
Sample Type DNA, dsDNA
Volume 16 x 1.25 mL
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.