Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega AccessQuick™ RT-PCR System

Catalog No. PRA1701 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
No. of Reactions:
100 Reactions
20 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PR-A17-01 20 Reactions
PRA1702 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-A17-01 Supplier Promega Supplier No. A1701
Add to Cart
Add to Cart

Includes

AccessQuick Master Mix, 2X; AMV reverse transcriptase; Nuclease-free water

The AccessQuick RT-PCR System is an easy and convenient master mix for one-tube RT-PCR.

  • Convenient RT-PCR in One Tube
  • Mix contains Tfl DNA Polymerase, dNTPs, magnesium sulfate and reaction buffer
  • Separate AMV RT provided for important no-RT control reactions
  • Simply combine AccessQuick™ Master Mix with AMV RT, RNA template, primers and water for efficient RT-PCR

For laboratory use.

Specifications

Content And Storage • 2X Master Mix, 2 x 250 μL
• Reverse Transcriptase, 1 x 100 μL
• Nuclease-free Water, 1 x 1.25 mL

Store at –30° to –10°C.
Kit Type AccessQuick™ RT-PCR system
No. of Tests 20 Tests
For Use With (Application) RT-PCR
No. of Reactions 20 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.