Learn More
Thermo Scientific™ Accucore™ 150 C18 HPLC Columns
Use this column, with its specially designed solid core, for high-resolution separation of biomolecules.
Specifications
For Use With | HPLC |
---|---|
Particle Size | 2.6 μm |
pH | 1 to 11 |
Pore Size | 150 Å |
Surface Area | 80 m²/g |
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
03-898-107
|
Thermo Scientific™
16126103030 |
2.6 μm | 3 mm | 100 mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
03-898-108
|
Thermo Scientific™
16126104630 |
2.6 μm | 4.6 mm | 100 mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
03-898-110
|
Thermo Scientific™
16126153030 |
2.6 μm | 3 mm | 150 mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
03-898-111
|
Thermo Scientific™
16126154630 |
2.6 μm | 4.6 mm | 150 mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
03-898-114
|
Thermo Scientific™
16126014005 |
2.6 μm | 4 mm | 10 mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
03-898-113
|
Thermo Scientific™
16126013005 |
2.6 μm | 3 mm | 10 mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
03-898-112
|
Thermo Scientific™
16126012105 |
2.6 μm | 2.1 mm | 10 mm | N/A | N/A | |||
This product is either new or inactive, and cannot be ordered online.Inactive Item. Please call Customer Service Representative. | |||||||||
03-898-106
|
Thermo Scientific™
16126102130 |
2.6 μm | 2.1 mm | 100 mm | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
03-898-109
|
Thermo Scientific™
16126152130 |
2.6 μm | 2.1 mm | 150 mm | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Offer phase characteristics designed for the separation of peptides with Thermo Scientific™ Accucore™ 150 C18 LC Columns. Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.
- 150Å pore size solid core particles for fast biomoleucule separations
- Superb resolution at low backpressures
- Exceptionally rugged columns
- Designed for the separation of peptides Outstanding resolution
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Specifications
HPLC | |
1 to 11 | |
80 m²/g | |
L1 | |
7% | |
Yes |
2.6 μm | |
150 Å | |
60°C | |
C18 | |
Reversed Phase | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.