Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Accucore™ 150-C4 HPLC Columns

Catalog No. 03898132
Click to view available options
Length (Metric):
10 mm
30 mm
50 mm
100 mm
150 mm
Diameter (Metric):
0.075 mm
2.1 mm
3 mm
4.6 mm

Use this specially designed solid core column with a C4 functionality for biomolecules that bind too tightly to C8 or C18 columns.

Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.

  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Significantly lower hydrophobic retention than C18
  • Ideal for optimal retention of proteins and larger peptides

Ordering Information:

Other custom column dimensions are available. Please call your local Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column, NanoViper
Diameter (Metric) 0.075 mm
For Use With HPLC
Length (Metric) 150 mm
Max. Pressure 11600 psi (800 bar)
Particle Size 2.6 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 80 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Product Line nanoLC
Packing Material Spherical, Solid Core, Ultrapure Silica
Carbon Load 2%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.