Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl CoA synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7246992 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166029 20 μg
NB166028 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166029 Supplier Novus Biologicals Supplier No. NBP31546620UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Acetyl CoA synthetase Polyclonal antibody specifically detects Acetyl CoA synthetase in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Acetyl CoA synthetase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias ACAS2, AceCS, acetate thiokinase, acetate-CoA ligase, Acetate--CoA ligase, Acetyl-CoA synthetase, acetyl-Coenzyme A synthetase 2 (ADP forming), acetyl-coenzyme A synthetase, cytoplasmic, ACSA, ACSACECS, Acyl-activating enzyme, acyl-CoA synthetase short-chain family member 2EC 6.2.1.1, cytoplasmic acetyl-coenzyme A synthetase, dJ1161H23.1, DKFZp762G026, EC 6.2.1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 367-606 of human Acetyl CoA synthetase (NP_001229322.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55902
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.